Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ism1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276489.1 | NP_001263418.1 | 2 | 825 | ||
GenBank mRNA | AK077398.1 | - | 2 | 826 |