Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177045.3 | NP_796019.1 | 20 | 2270 | ||
GenBank mRNA | AK043839.1 | - | 20 | 2271 | ||
AK146601.1 | - | 20 | 2289 | |||
AK173271.1 | - | 16 | 1698 | |||
BC055068.1 | - | 18 | 1912 | |||
BC062883.1 | - | 11 | 1893 | |||
BC125517.1 | - | 19 | 2160 | 40130032 | ||
BC137743.1 | - | 19 | 2160 | 8860762 |