Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRF SIRNA Assays ›
Gene Symbol:
PTPRF
Gene Name:
protein tyrosine phosphatase, receptor type F
Gene Aliases:
BNAH2, LAR
Chromosome Location:
Chr.1: 43522238 - 43623672 on Build GRCh38
UniGene ID:
Hs.272062
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s11546
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005271079.3
XP_005271136.1
16
2757
XM_005271080.3
XP_005271137.1
15
2745
XM_005271081.3
XP_005271138.1
14
2718
XM_005271082.3
XP_005271139.1
13
2412
XM_006710795.3
XP_006710858.1
17
2735
XM_006710796.3
XP_006710859.1
15
2730
XM_006710797.3
XP_006710860.1
16
2757
XM_006710798.3
XP_006710861.1
15
2730
XM_006710799.3
XP_006710862.1
15
2451
XM_006710800.3
XP_006710863.1
14
2424
XM_006710801.3
XP_006710864.1
16
2757
XM_011541871.2
XP_011540173.1
17
2769
XM_011541872.2
XP_011540174.1
17
2755
XM_011541873.2
XP_011540175.1
17
2689
XM_017001942.1
XP_016857431.1
14
2386
XM_017001943.1
XP_016857432.1
14
2439
XM_017001944.1
XP_016857433.1
14
2424
XM_017001945.1
XP_016857434.1
13
2412
XM_017001946.1
XP_016857435.1
15
2451
XM_017001947.1
XP_016857436.1
14
2424
XM_017001948.1
XP_016857437.1
14
2450
XM_017001949.1
XP_016857438.1
15
2415
XM_017001950.1
XP_016857439.1
13
2420
GenBank mRNA
AB177856.1
-
15
1948
AB177857.1
-
14
2677
AB209478.1
-
6
1332
AK125325.1
-
5
629
AK127007.1
-
3
151
BC048768.1
-
14
2688
5502598
BX537361.1
-
2
889
Y00815.1
-
15
2729
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00160858_m1
Panther Classification:
Molecular Function -
protein phosphatase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
cell migration
neuron projection regeneration
peptidyl-tyrosine dephosphorylation
regulation of axon regeneration
negative regulation of receptor binding
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
heparin binding
protein complex binding
chondroitin sulfate proteoglycan binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-nbn7j:80/100.66.77.222:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline