Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198899.2 | NP_942602.2 | 28, 29 | 3288 | ||
GenBank mRNA | AK144381.1 | - | 27, 28 | 3117 | ||
BC023910.1 | - | 1, 2 | 84 | |||
BC051172.1 | - | 2, 3 | 392 | |||
BC062936.1 | - | 28, 29 | 3235 | 6838545 | ||
CD808359.1 | - | 1, 2 | 51 |