Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198899.2 | NP_942602.2 | 14 | 1581 | ||
GenBank mRNA | AK050532.1 | - | 14 | 1568 | ||
AK087883.1 | - | 14 | 1380 | |||
AK144381.1 | - | 14 | 1434 | |||
BC062936.1 | - | 14 | 1528 | 6838545 | ||
BC068283.1 | - | 14 | 1563 | 30022247 |