Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 6 | 961 | ||
GenBank mRNA | AK036709.1 | - | 6 | 962 | ||
AK046672.1 | - | 962 | ||||
AK133242.1 | - | 6 | 1822 | |||
AK155881.1 | - | 6 | 977 | |||
AK170621.1 | - | 6 | 983 | |||
AK171230.1 | - | 6 | 2640 | |||
AK171587.1 | - | 6 | 962 | |||
AK171603.1 | - | 6 | 960 | |||
AK171605.1 | - | 6 | 977 | |||
AY522648.1 | - | 6 | 962 | |||
DQ087179.1 | - | 6 | 714 | |||
DQ087180.1 | - | 7 | 805 |