Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 2 | 593 | ||
GenBank mRNA | AK036709.1 | - | 2 | 594 | ||
AK046672.1 | - | 594 | ||||
AK133242.1 | - | 2 | 1454 | |||
AK155881.1 | - | 2 | 609 | |||
AK157919.1 | - | 2 | 332 | |||
AK170621.1 | - | 2 | 615 | |||
AK171230.1 | - | 2 | 592 | |||
AK171587.1 | - | 2 | 594 | |||
AK171603.1 | - | 2 | 592 | |||
AK171605.1 | - | 2 | 609 | |||
AY522648.1 | - | 2 | 594 | |||
DQ087179.1 | - | 2 | 346 | |||
DQ087180.1 | - | 2 | 346 |