Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 3 | 347 | ||
GenBank mRNA | AK038670.1 | - | 3 | 291 | ||
AK038681.1 | - | 3 | 291 | |||
AK039575.1 | - | 3 | 348 | |||
AK046891.1 | - | 3 | 289 | |||
AK122406.1 | - | 3 | 421 | |||
AK134967.1 | - | 3 | 296 | |||
AK162922.1 | - | 3 | 285 | |||
BC027101.1 | - | 3 | 276 | |||
BC058772.1 | - | 3 | 288 | |||
BC080664.1 | - | 3 | 286 | |||
BC117788.1 | - | 3 | 289 | 40092137 | ||
BC117789.2 | - | 3 | 289 | 40092140 |