Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172554.2 | NP_766142.2 | 5 | 553 | ||
GenBank mRNA | AK038670.1 | - | 5 | 497 | ||
AK038681.1 | - | 5 | 497 | |||
AK039575.1 | - | 5 | 554 | |||
AK046891.1 | - | 5 | 495 | |||
AK122406.1 | - | 5 | 627 | |||
AK134967.1 | - | 5 | 502 | |||
AK162922.1 | - | 5 | 491 | |||
BC027101.1 | - | 5 | 482 | |||
BC058772.1 | - | 5 | 494 | |||
BC080664.1 | - | 5 | 492 | |||
BC117788.1 | - | 5 | 495 | 40092137 | ||
BC117789.2 | - | 5 | 495 | 40092140 |