Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002843.3 | NP_002834.3 | 9 | 2117 | ||
XM_017018083.1 | XP_016873572.1 | 8 | 1848 | |||
XM_017018084.1 | XP_016873573.1 | 9 | 2230 | |||
XM_017018085.1 | XP_016873574.1 | 9 | 1729 | |||
GenBank mRNA | D37781.1 | - | 9 | 2106 | ||
U10886.1 | - | 9 | 2111 |