Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135648.2 | NP_001129120.1 | 6 | 1039 | ||
NM_001291981.1 | NP_001278910.1 | 6 | 1039 | |||
NM_001291982.1 | NP_001278911.1 | 6 | 1039 | |||
NM_001291983.1 | NP_001278912.1 | 5 | 767 | |||
NM_001291984.1 | NP_001278913.1 | 6 | 1039 | |||
NM_002844.3 | NP_002835.2 | 6 | 1039 | |||
XM_006715537.2 | XP_006715600.1 | 6 | 1090 | |||
XM_011536014.2 | XP_011534316.1 | 6 | 1090 | |||
XM_011536015.2 | XP_011534317.1 | 6 | 1090 | |||
XM_011536016.2 | XP_011534318.1 | 6 | 732 | |||
XM_011536017.2 | XP_011534319.1 | 7 | 1330 | |||
XM_011536018.2 | XP_011534320.1 | 7 | 1003 | |||
XM_011536020.2 | XP_011534322.1 | 6 | 1090 | |||
XM_011536021.2 | XP_011534323.1 | 6 | 1090 | |||
XM_017011145.1 | XP_016866634.1 | 7 | 1054 | |||
XM_017011146.1 | XP_016866635.1 | 7 | 1003 | |||
XM_017011147.1 | XP_016866636.1 | 7 | 984 | |||
XM_017011148.1 | XP_016866637.1 | 7 | 981 | |||
GenBank mRNA | AB209670.1 | - | 4 | 280 | ||
AF533875.1 | - | 6 | 804 | |||
AK295028.1 | - | 6 | 964 | |||
BC140775.1 | - | 6 | 841 | 9021643 | ||
BC144512.1 | - | 6 | 841 | 9053039 | ||
BC144513.1 | - | 6 | 841 | 9053040 | ||
CR749277.1 | - | 6 | 980 | |||
L77886.1 | - | 6 | 738 | |||
Z70660.1 | - | 6 | 723 |