Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177320.2 | NP_796294.2 | 9, 10 | 1049 | ||
GenBank mRNA | AK029201.1 | - | 7, 8 | 717 | ||
AK089483.1 | - | 9, 10 | 1054 | |||
AK149847.1 | - | 9, 10 | 1017 | |||
AK154643.1 | - | 9, 10 | 1028 | |||
AK171279.1 | - | 9, 10 | 1050 | |||
AY156924.1 | - | 8, 9 | 898 | |||
BC128076.1 | - | 8, 9 | 898 | 40060132 | ||
BC128077.1 | - | 8, 9 | 898 | 40060133 |