Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177320.2 | NP_796294.2 | 17 | 2486 | ||
GenBank mRNA | AK149847.1 | - | 17 | 2454 | ||
AK154643.1 | - | 17 | 2465 | |||
AK161050.1 | - | 8 | 1394 | |||
AY156924.1 | - | 16 | 2335 | |||
BC128076.1 | - | 16 | 2335 | 40060132 | ||
BC128077.1 | - | 16 | 2335 | 40060133 |