Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177103.4 | NP_796077.2 | 2 | 1033 | ||
GenBank mRNA | AK028177.1 | - | 2 | 1034 | ||
AK042087.1 | - | 2 | 925 | |||
AK043171.1 | - | 2 | 918 | |||
AK136120.1 | - | 1 | 626 | |||
AK158338.1 | - | 2 | 898 | |||
AY522918.1 | - | 2 | 897 | |||
BC067014.1 | - | 2 | 800 | 6812169 | ||
BC080830.1 | - | 2 | 917 | 30433573 |