Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177111.3 | NP_796085.3 | 16 | 2431 | ||
GenBank mRNA | AK087583.1 | - | 8 | 1152 | ||
AK158666.1 | - | 14 | 1949 | |||
AK170006.1 | - | 14 | 1898 | |||
BC070471.1 | - | 16 | 2432 | 30626499 | ||
BC072583.1 | - | 8 | 1548 | 6403859 |