Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081423.2 | NP_001074892.2 | 7 | 763 | ||
GenBank mRNA | AB093278.1 | - | 7 | 696 | ||
AK029125.1 | - | 6 | 2408 | |||
AK085566.1 | - | 7 | 1007 | |||
AM690748.1 | - | 6 | 528 | |||
BC035276.1 | - | 6 | 638 |