Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145839.2 | NP_665838.1 | 7 | 1026 | ||
NM_181318.4 | NP_851835.1 | 7 | 1023 | |||
GenBank mRNA | AK028756.1 | - | 7 | 915 | ||
AK090085.1 | - | 7 | 918 | |||
AK149812.1 | - | 7 | 945 | |||
AY129963.1 | - | 7 | 953 | |||
BC050858.1 | - | 7 | 956 | 6517502 | ||
BC052726.1 | - | 7 | 917 | 6813357 |