Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145839.2 | NP_665838.1 | 3 | 426 | ||
NM_181318.4 | NP_851835.1 | 3 | 426 | |||
GenBank mRNA | AK028756.1 | - | 3 | 318 | ||
AK090085.1 | - | 3 | 318 | |||
AK149812.1 | - | 3 | 345 | |||
AY129963.1 | - | 3 | 353 | |||
BC050858.1 | - | 3 | 359 | 6517502 | ||
BC052726.1 | - | 3 | 317 | 6813357 |