Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D830030K20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177135.4 | NP_796109.1 | 4 | 454 | ||
GenBank mRNA | AK082998.1 | - | 4 | 531 | ||
AK085928.1 | - | 4 | 455 | |||
EF651825.1 | - | 3 | 311 |
Gene Symbol: Gm10408 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256501.1 | NP_001243430.1 | 4 | 596 |