Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177161.4 | NP_796135.3 | 1, 2 | 204 | ||
GenBank mRNA | AK086780.1 | - | 1, 2 | 185 | ||
AY313449.1 | - | 1, 2 | 204 |