Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207015.1 | NP_001193944.1 | 8 | 1098 | ||
NM_001207016.1 | NP_001193945.1 | 6 | 888 | |||
NM_002849.3 | NP_002840.2 | 9 | 1754 | |||
NM_130846.2 | NP_570897.2 | 5 | 1020 | |||
NR_073474.1 | 4 | 833 | ||||
XM_011538615.2 | XP_011536917.1 | 9 | 1350 | |||
GenBank mRNA | AK295951.1 | - | 8 | 1088 | ||
AK304457.1 | - | 6 | 847 | |||
AK304672.1 | - | 6 | 888 | |||
AK312376.1 | - | 9 | 1667 | |||
BC110900.1 | - | 5 | 717 | 4538615 | ||
BC119777.1 | - | 6 | 696 | 40109494 | ||
BC122866.1 | - | 6 | 696 | 40109492 | ||
BX571751.1 | - | 5 | 1073 | |||
D64053.1 | - | 9 | 1756 | |||
U42361.1 | - | 9 | 1514 | |||
U77916.1 | - | 6 | 738 | |||
U77917.1 | - | 5 | 997 | |||
X82635.1 | - | 5 | 978 | |||
Z79693.1 | - | 9 | 1425 |