Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177171.4 | NP_796145.2 | 24 | 3846 | ||
GenBank mRNA | AK053055.1 | - | 24 | 3846 | ||
AK122493.1 | - | 13 | 1850 | |||
AK161444.1 | - | 13 | 2164 | |||
BC086656.1 | - | 23 | 3671 | 30544145 |