Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207015.1 | NP_001193944.1 | 10 | 1353 | ||
NM_001207016.1 | NP_001193945.1 | 8 | 1143 | |||
NM_002849.3 | NP_002840.2 | 11 | 2009 | |||
NM_130846.2 | NP_570897.2 | 7 | 1275 | |||
NR_073474.1 | 7 | 1345 | ||||
XM_011538615.2 | XP_011536917.1 | 11 | 1605 | |||
GenBank mRNA | AK091647.1 | - | 2 | 138 | ||
AK295951.1 | - | 10 | 1343 | |||
AK304457.1 | - | 8 | 1102 | |||
AK304672.1 | - | 8 | 1143 | |||
AK312376.1 | - | 11 | 1922 | |||
BC110900.1 | - | 7 | 972 | 4538615 | ||
BC119777.1 | - | 8 | 951 | 40109494 | ||
BC122866.1 | - | 8 | 951 | 40109492 | ||
BX571751.1 | - | 7 | 1328 | |||
CR749836.1 | - | 2 | 408 | |||
D64053.1 | - | 11 | 2011 | |||
U42361.1 | - | 11 | 1769 | |||
U77916.1 | - | 8 | 993 | |||
U77917.1 | - | 7 | 1252 | |||
X82635.1 | - | 7 | 1233 | |||
Z79693.1 | - | 11 | 1680 |