Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A830018L16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160369.2 | NP_001153841.1 | 2 | 670 | ||
NM_001160370.2 | NP_001153842.1 | 2 | 670 | |||
NM_001160371.2 | NP_001153843.1 | 2 | 670 | |||
NM_177173.6 | NP_796147.2 | 2 | 670 | |||
GenBank mRNA | AK034391.1 | - | 2 | 666 | ||
AK039555.1 | - | 2 | 477 | |||
AK043673.1 | - | 2 | 507 | |||
AK081622.1 | - | 2 | 671 | |||
AK135047.1 | - | 2 | 664 | |||
AK136763.1 | - | 2 | 471 | |||
BC137903.1 | - | 2 | 266 | 8860922 | ||
BC145022.1 | - | 2 | 266 | 9053570 | ||
BC145023.1 | - | 2 | 266 | 9053571 |