Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000317.2 | NP_000308.1 | 4 | 287 | ||
GenBank mRNA | AK310768.1 | - | 3 | 241 | ||
BC009686.1 | - | 4 | 251 | 3901665 | ||
BC018029.1 | - | 4 | 283 | 4819386 | ||
D17400.1 | - | 4 | 276 | |||
M97655.1 | - | 4 | 217 |