Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177222.4 | NP_796196.3 | 14, 15 | 1960 | ||
GenBank mRNA | AY423542.1 | - | 14, 15 | 1891 | ||
AY485607.1 | - | 14, 15 | 1960 | |||
AY485608.1 | - | 14, 15 | 1960 | |||
BC120743.1 | - | 14, 15 | 1864 | 40129666 | ||
BC125423.1 | - | 14, 15 | 1864 | 40129938 | ||
BC144801.1 | - | 14, 15 | 1864 |