Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dctd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161515.1 | NP_001154987.1 | 4 | 606 | ||
NM_001161516.1 | NP_001154988.1 | 4 | 387 | |||
NM_178788.4 | NP_848903.2 | 4 | 628 | |||
GenBank mRNA | AK031645.1 | - | 4 | 593 | ||
AK039066.1 | - | 4 | 418 | |||
AK077663.1 | - | 4 | 590 | |||
AK138614.1 | - | 4 | 416 | |||
AK171894.1 | - | 4 | 425 | |||
BB193146.2 | - | 2 | 197 | |||
BC031719.1 | - | 4 | 412 | 3672932 | ||
BQ572929.1 | - | 2 | 102 | |||
CO803572.1 | - | 4 | 387 |