Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002852.3 | NP_002843.2 | 3 | 1044 | ||
GenBank mRNA | AK312705.1 | - | 3 | 1044 | ||
BC039733.1 | - | 3 | 1017 | 5768379 | ||
M31166.1 | - | 3 | 974 | |||
X63053.1 | - | 3 | 906 | |||
X63613.1 | - | 3 | 968 |