Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178789.4 | NP_848904.1 | 4, 5 | 658 | ||
GenBank mRNA | AK047437.1 | - | 4, 5 | 659 | ||
AK048597.1 | - | 4, 5 | 618 | |||
AK052935.1 | - | 2, 3 | 250 | |||
BC051178.1 | - | 4, 5 | 593 | 6314268 | ||
BC054070.1 | - | 4, 5 | 593 |