Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mysm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177239.2 | NP_796213.2 | 17, 18 | 2173 | ||
GenBank mRNA | AK173300.1 | - | 14, 15 | 1915 | ||
BC150946.1 | - | 17, 18 | 2169 | 9087857 | ||
BC151172.1 | - | 17, 18 | 2169 | 9088061 |