Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177263.3 | NP_796237.2 | 3 | 1446 | ||
GenBank mRNA | AB099527.2 | - | 1 | 990 | ||
AK029603.1 | - | 3 | 1318 | |||
AK030057.1 | - | 3 | 1411 | |||
AK143296.1 | - | 3 | 1427 | |||
AK147259.1 | - | 1266 | ||||
BC058111.1 | - | 3 | 1446 | 6836697 | ||
DQ317973.1 | - | 1 | 990 |