Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033397.5 | NP_001028569.2 | 1, 2 | 496 | ||
GenBank mRNA | AK028591.1 | - | 1, 2 | 496 | ||
AK028682.1 | - | 1, 2 | 496 | |||
AK132445.1 | - | 1, 2 | 500 | |||
AK162718.1 | - | 1, 2 | 498 | |||
BC116672.1 | - | 1, 2 | 485 | 40107596 |