Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193305.1 | NP_001180234.1 | 4, 5 | 801 | ||
NM_177282.5 | NP_796256.1 | 4, 5 | 801 | |||
GenBank mRNA | AK030608.1 | - | 4, 5 | 782 | ||
AK138935.1 | - | 4, 5 | 775 | |||
AK143191.1 | - | 4, 5 | 748 | |||
AK161075.1 | - | 4, 5 | 748 | |||
AK163078.1 | - | 4, 5 | 768 | |||
AK163231.1 | - | 4, 5 | 747 | |||
AK220353.1 | - | 2, 3 | 478 | |||
BC111895.1 | - | 2, 3 | 525 | 40061129 | ||
BC112415.1 | - | 2, 3 | 525 | 40061128 |