Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177290.3 | NP_796264.2 | 12 | 2481 | ||
GenBank mRNA | AK082444.1 | - | 3073 | |||
AK140969.1 | - | 3 | 569 | |||
BC120691.1 | - | 12 | 2481 | 40129614 | ||
BC125343.1 | - | 12 | 2481 | 40129858 | ||
BC144785.1 | - | 12 | 2486 |