Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000027.3 | NP_000018.2 | 9 | 1123 | ||
NM_001171988.1 | NP_001165459.1 | 9 | 1093 | |||
NR_033655.1 | 8 | 1047 | ||||
GenBank mRNA | AK312982.1 | - | 9 | 1057 | ||
BC012392.1 | - | 9 | 1046 | 3859317 | ||
CR541715.1 | - | 9 | 995 | |||
M64073.1 | - | 9 | 998 | |||
X55762.1 | - | 9 | 1022 |