Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pisd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177298.3 | NP_796272.2 | 7 | 900 | ||
GenBank mRNA | AK033483.1 | - | 7 | 892 | ||
AK034656.1 | - | 7 | 902 | |||
AK162028.1 | - | 7 | 893 | |||
AK162737.1 | - | 7 | 901 | |||
AK167554.1 | - | 6 | 842 | |||
BC070408.1 | - | 7 | 899 | 6809528 | ||
BC094594.1 | - | 7 | 891 | 5707749 | ||
BC116695.1 | - | 7 | 902 | 40105525 | ||
BC117509.1 | - | 7 | 902 | 40105529 |
Gene Symbol: Pisd-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CJ124393.1 | - | 2 | 89 |
Gene Symbol: Pisd-ps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034517.1 | - | 7 | 1721 | ||
AK036319.1 | - | 7 | 1346 | |||
AK081907.1 | - | 5 | 1546 | |||
AK144226.1 | - | 3 | 1060 | |||
AK161130.1 | - | 7 | 1111 | |||
AK162734.1 | - | 6 | 2460 | |||
AK168892.1 | - | 6 | 1304 | |||
AK169597.1 | - | 4 | 2311 | |||
AK169755.1 | - | 4 | 2311 | |||
BC003217.1 | - | 1 | 181 | |||
BC034366.1 | - | 2 | 2079 | |||
BC048401.1 | - | 5 | 975 | |||
BC059938.1 | - | 6 | 970 | |||
BC083061.1 | - | 6 | 812 |