Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177303.4 | NP_796277.2 | 5 | 1361 | ||
GenBank mRNA | AK080889.1 | - | 5 | 1362 | ||
AK085246.1 | - | 5 | 1391 | |||
AY325108.1 | - | 5 | 1350 |