Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134426.1 | NP_001127898.1 | 2 | 771 | ||
NM_001134427.1 | NP_001127899.1 | 2 | 561 | |||
NM_175565.3 | NP_780774.1 | 2 | 771 | |||
NM_175833.2 | NP_787027.1 | 2 | 771 | |||
GenBank mRNA | AB046372.1 | - | 2 | 359 | ||
AF215660.1 | - | 2 | 529 | |||
AF320340.1 | - | 2 | 529 | |||
AK010956.1 | - | 2 | 663 | |||
AK048941.1 | - | 2 | 955 | |||
AK172053.1 | - | 2 | 480 | |||
BC031362.1 | - | 2 | 561 | |||
BC052770.1 | - | 2 | 665 | 6306557 | ||
BC062156.1 | - | 3 | 617 | 6507616 | ||
BC096479.1 | - | 2 | 622 | 4979176 | ||
L22067.1 | - | 2 | 612 |