Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177794.3 | NP_808462.2 | 6 | 1107 | ||
GenBank mRNA | AK028283.1 | - | 6 | 1108 | ||
AK084351.1 | - | 6 | 1085 | |||
AK143866.1 | - | 6 | 1085 | |||
BC117502.1 | - | 7 | 1111 | 40106131 | ||
BC117503.1 | - | 6 | 1029 | 40106128 |