Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205341.1 | NP_001192270.1 | 28 | 3661 | ||
NM_177373.4 | NP_796347.2 | 28 | 3658 | |||
GenBank mRNA | AK087448.1 | - | 7 | 1157 | ||
AK220491.1 | - | 28 | 3618 | |||
BC065133.1 | - | 28 | 3629 | 30536236 | ||
EU568870.1 | - | 28 | 3632 |