Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000027.3 | NP_000018.2 | 3 | 443 | ||
NM_001171988.1 | NP_001165459.1 | 3 | 443 | |||
NR_033655.1 | 3 | 443 | ||||
GenBank mRNA | AK312982.1 | - | 3 | 377 | ||
BC012392.1 | - | 3 | 366 | 3859317 | ||
CR541715.1 | - | 3 | 315 | |||
CX869892.1 | - | 3 | 320 | |||
DA346002.1 | - | 3 | 443 | |||
DA905583.1 | - | 3 | 433 | |||
M64073.1 | - | 3 | 318 | |||
M64075.1 | - | 11 | ||||
M64076.1 | - | 11 | ||||
X55762.1 | - | 3 | 342 |