Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024927.2 | NP_001020098.1 | 18 | 2633 | ||
NM_001081641.1 | NP_001075110.1 | 17 | 2585 | |||
GenBank mRNA | AK147444.1 | - | 18 | 2590 | ||
AK147630.1 | - | 17 | 2418 | |||
BC112411.1 | - | 14 | 2547 | 40061037 | ||
CF531644.1 | - | 1 | 50 |