Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177815.3 | NP_808483.2 | 12 | 1485 | ||
GenBank mRNA | BC082539.1 | - | 12 | 1460 | ||
BC094336.1 | - | 11 | 1347 | |||
BC141275.1 | - | 12 | 1479 | 9055887 | ||
BC145606.1 | - | 12 | 1392 | 9054172 |