Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005050.3 | NP_005041.1 | 3 | 383 | ||
XM_005267939.3 | XP_005267996.1 | 3 | 316 | |||
XM_005267940.3 | XP_005267997.1 | 2 | 195 | |||
XM_005267949.3 | XP_005268006.1 | 3 | 381 | |||
XM_006720223.2 | XP_006720286.1 | 2 | 195 | |||
XM_017021531.1 | XP_016877020.1 | 3 | 316 | |||
XM_017021532.1 | XP_016877021.1 | 3 | 316 | |||
XM_017021533.1 | XP_016877022.1 | 3 | 316 | |||
XM_017021534.1 | XP_016877023.1 | 2 | 142 | |||
XM_017021535.1 | XP_016877024.1 | 3 | 316 | |||
XM_017021538.1 | XP_016877027.1 | 2 | 142 | |||
XM_017021540.1 | XP_016877029.1 | 2 | 129 | |||
XM_017021541.1 | XP_016877030.1 | 3 | 253 | |||
XM_017021542.1 | XP_016877031.1 | 2 | 159 | |||
XM_017021543.1 | XP_016877032.1 | 2 | 142 | |||
XM_017021544.1 | XP_016877033.1 | 3 | 316 | |||
GenBank mRNA | AF009746.1 | - | 3 | 282 | ||
AK291332.1 | - | 3 | 253 | |||
AK297934.1 | - | 2 | 169 | |||
BC012815.2 | - | 3 | 237 | 2960427 | ||
BI916327.1 | - | 3 | 375 | |||
BT007412.1 | - | 3 | 231 | |||
BX247984.1 | - | 2 | 192 | |||
CR457104.1 | - | 3 | 231 | |||
DC332064.1 | - | 3 | 383 |