Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177815.3 | NP_808483.2 | 8, 9 | 875 | ||
GenBank mRNA | AK184779.1 | - | 3, 4 | 276 | ||
BC082539.1 | - | 8, 9 | 850 | |||
BC094336.1 | - | 8, 9 | 843 | |||
BC141275.1 | - | 8, 9 | 869 | 9055887 |