Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000027.3 | NP_000018.2 | 6 | 801 | ||
GenBank mRNA | AK312982.1 | - | 6 | 735 | ||
BC012392.1 | - | 6 | 724 | 3859317 | ||
CR541715.1 | - | 6 | 673 | |||
M64073.1 | - | 6 | 676 | |||
X55762.1 | - | 6 | 700 |