Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ep300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177821.6 | NP_808489.4 | 18 | 3802 | ||
GenBank mRNA | AK139715.1 | - | 15 | 2296 | ||
BC144976.1 | - | 18 | 3656 | 9053524 | ||
BC150681.1 | - | 18 | 3656 | 9087592 | ||
BY759831.1 | - | 3 | 393 | |||
CB245766.1 | - | 2 | 224 | |||
CN535246.1 | - | 3 | 320 | |||
CN537190.1 | - | 4 | 507 |