Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm813 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033404.2 | NP_001028576.1 | 3 | 327 | ||
GenBank mRNA | AK136264.1 | - | 3 | 328 | ||
BC145581.1 | - | 3 | 297 | |||
BC145922.1 | - | 3 | 326 | 40131225 | ||
BC145924.1 | - | 3 | 326 | 40131227 |