Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177823.4 | NP_808491.2 | 6, 7 | 993 | ||
GenBank mRNA | AK040572.1 | - | 6, 7 | 954 | ||
AK041690.1 | - | 6, 7 | 960 | |||
AK049106.1 | - | 5, 6 | 893 |