Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289497.1 | NP_001276426.1 | 3 | 509 | ||
NM_177834.4 | NP_808502.2 | 4 | 634 | |||
GenBank mRNA | AK078883.1 | - | 3 | 510 | ||
AY773477.1 | - | 4 | 632 | |||
BC119559.1 | - | 4 | 619 | 40057538 | ||
BC119560.1 | - | 4 | 619 | 40057539 |